Thermo Scientific™ HotStart™ Storage Reaction Tubes Available on GSA/VA Contract for Federal Government customers only.

Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.

Manufacturer: Thermo Scientific™ 630211

 View more versions of this product

Catalog No. 21-236-62

  Check Availability
Add to Cart

    Specifications

    For Use With (Equipment) Thermal Cyclers
    Tube Type Individual
    Reaction Volume 25 to 50µL
    Type Tubes
    Volume (Metric) 0.5mL
    Description Hot Start Micro 50 Bulk

    Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.

    Features:

    • Bead is stable at room temperature
    • Assures synchronous reaction startup
    • PCR product can remain in tube for later analysis
    • Hinged caps provide snug seal
    • Presterilized; RNase- and DNase-free
    Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit