Learn More
Thermo Scientific™ HotStart™ Storage Reaction Tubes
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Manufacturer:  Thermo Scientific™ 631011
 View more versions of this product
Catalog No. 21-236-65
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
Specifications
Specifications
0.2 mL
Tubes
25 to 50µL
Thermal Cyclers
Sterile
Hot Start Micro 50 Bulk
0.2 mL
Individual