Learn More
Thermo Scientific™ HotStart™ Storage Reaction Tubes
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Specifications
For Use With (Equipment) Thermal Cyclers
View More Specs
Products 2
Catalog Number Mfr. No. Description Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Description Volume (Metric) Price Quantity & Availability  
21-236-65
View Documents
Thermo Scientific™
631011
Hot Start Micro 50 Bulk 0.2 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
21-236-61
View Documents
Thermo Scientific™
6210
HotStart Micro 50 Strips 0.2 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
Eliminate tedious oil or wax overlays with the thin-walled Thermo Scientific™ HotStart™ Storage Reaction Tubes.
Features:
  • Bead is stable at room temperature
  • Assures synchronous reaction startup
  • PCR product can remain in tube for later analysis
  • Hinged caps provide snug seal
  • Presterilized; RNase- and DNase-free
Recommended for: Complex DNA or cDNA, very low copy number targets, more than 30 thermal cycles, multiple PCR or RT-PCR
Specifications
Specifications
Thermal Cyclers