Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International™ HandPRO™ AQUA1900™ Nitrile Gloves
BIOPHARMA

Catalog Number p-4390824
Click to view available options
Size:
Small
Medium
Large
X-Large

Powder free, general purpose nitrile gloves are suitable for veterinary, animal lab, industrial or other non-medical applications.

  • Powder free
  • Not made with natural rubber latex
  • Textured fingertips for enhanced grip and tactile sensitivity
  • Single chlorination for donning ease
  • Beaded cuff
  • Static dissipative ESD qualities
  • Ocean blue color
Certifications/Compliance Meets or exceeds ASTM requirements
Accelerator Free No
Applications Industrial
Color Blue
Exterior Finish Partially Textured
Finger Thickness (English) 5.5 mil
Finger Thickness (Metric) 0.14 mm
Latex Free Yes
Material Nitrile
Palm Thickness (English) 3.1 mil
Palm Thickness (Metric) 0.08 mm
Powdered Powder Free
Size Small
Thickness (English) 5.5 mil
Thickness (Metric) 0.14 mm
Model 1901
Tensile Strength 43 Mpa
Length (English) 9 in.
Length (Metric) 23 cm
Show More Show Less
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit