Learn More
Hourglass International™ HandPRO™ LSCT 400S Latex Sterile Surgical Gloves
Smooth and tapered
Manufacturer:  Hourglass International™ LSCT4090S
 View more versions of this product
Catalog No. 180-401-260
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Alert:
These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.
Description
Description
  • Powder-free
  • With beaded cuff
  • Chlorinated
  • Hand specific
  • Gamma sterilized
  • Natural color
Compliance
Meet ASTM™ D3588-91/D3578-91 standards.
Specifications
Specifications
Beaded/Rolled
9
Natural
16 in.
Latex
Powder-free
Sterile
40cm