Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hourglass International™ HandPRO™ LSCT 400S Latex Sterile Surgical Gloves

Sterile, powder free latex gloves are smooth and tapered for precise instrument handling.

Specifications

Color Natural
Length (English) 16 in.
Length (Metric) 40 cm
Material Latex
Sterility Sterile
View More Specs

Alert:

These products contain natural rubber latex, a component of which may cause allergic reaction in some users. Use only as directed and in accordance with manufacturer instructions and warnings. Please consult your institution's policies regarding use of these products.


Products 6
Catalog Number Mfr. No. Size Price Quantity & Availability  
Catalog Number Mfr. No. Size Price Quantity & Availability  
180-401-257
View Documents
Hourglass International™
LSCT4075S
7.5 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
180-401-258
View Documents
Hourglass International™
LSCT4080S
8 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
180-401-259
View Documents
Hourglass International™
LSCT4085S
8.5 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
180-401-260
View Documents
Hourglass International™
LSCT4090S
9 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
180-401-261
View Documents
Hourglass International™
LSCT4095S
9.5 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
180-401-256
View Documents
Hourglass International™
LSCT4010S
10 This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.Due to product restrictions, please to purchase or view availability for this product. N/A
Description

Description

  • Powder free
  • With beaded cuff
  • Chlorinated
  • Hand specific
  • Gamma sterilized
  • Natural color

Specifications

Specifications

Natural
40 cm
Sterile
16 in.
Latex
Powder Free
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit