Learn More
SiliCycle™ Housing Kit for Laboratory Scale E-PAK™ Cartridges
This Housing Kit is for Laboratory Scale E-PAK™ Cartridges, which are generally used for processing samples of 50 mL or more.
Supplier:  SiliCycle™ LSHK25S
 View more versions of this product
Catalog No. 01-182-151
Add to cart
Description
Description
  • 5 x 1 cm
  • 5 x 10 cm
  • 5 x 25 cm
  • Kits including bowls of various sizes
Specifications
Specifications
Lab Scale E-PAK Housing
1 Bowl (5 x 25 cm)
1 Kit
5 x 25 cm Laboratory Scale E-PAK™ Cartridges