Learn More
How Viruses Travel Lab Investigation Products for Science Education
Simulate the spread of disease using a safe, simulated disease agent.
Manufacturer:  Neo/Sci Corporation 201135
 View more versions of this product
Catalog No. S32009
This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes:
  • Simulated virus and virus detection agent
  • Test vials
  • Micropipets
  • Microcentrifuge tubes
  • Comprehensive teacher and student guides
Description
Description
  • Model the transmission of a communicable viral disease, identify its origin and calculate its rate of spread through the population
  • Allows class to form inferences on the rate at which a disease spreads today as compared to a similar outbreak a century ago
  • Class size: 40 students
Specifications
Specifications
Simulate the spread of disease using a safe, simulated disease agent
Refill
How Viruses Travel Refill Kit