Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

How Viruses Travel Lab Investigation Products for Science Education

Simulate the spread of disease using a safe, simulated disease agent.

Specifications

For Use With (Application) Simulate the spread of disease using a safe, simulated disease agent
View More Specs

Includes:
  • Simulated virus and virus detection agent
  • Test vials
  • Micropipets
  • Microcentrifuge tubes
  • Comprehensive teacher and student guides

Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
S32008 Products for Science Education
View Documents
Neo/Sci Corporation
201133
How Viruses Travel Lab Investigation This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
S32009 Products for Science Education
View Documents
Neo/Sci Corporation
201135
How Viruses Travel Refill Kit This item has been discontinued and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Model the transmission of a communicable viral disease, identify its origin and calculate its rate of spread through the population
  • Allows class to form inferences on the rate at which a disease spreads today as compared to a similar outbreak a century ago
  • Class size: 40 students

Specifications

Specifications

Simulate the spread of disease using a safe, simulated disease agent
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit