Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Howard Leight™ AirSoft™ Ear Plugs

NRR: 27dB

Manufacturer:  Honeywell Safety Products DPAS30W

 View more versions of this product

Catalog No. 18-534



  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

4.87 in.
Blue
Airsoft™ Earplug
TPE
4 Flange
White Nylon cord, Packed in Flip-Top Box
27dB
4.87 x 5.5 x 8.12 in. (12.3 x 3.8 x 20.6cm)
12.3cm
Four flange design for better seal in ear canal, optimal protection, and increased comfort for long-term wear
8.12 in.
20.6cm
3.8cm
5.5 in.
ANSI S3.19-1974
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit