Howard Leight™ Clarity™ C1 Earmuffs
Provide uniform attenuation that allows speech frequencies to be heard with less distortion
Manufacturer: Honeywell Safety Products 1011142
 View more versions of this product
Catalog No. 19-161-267
  Check Availability
Add to Cart
    Specifications
    For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, Metal fabrication, petrochemical Steel
    Material Plastic
    Style Over-the-head
    Product Type Headband Ear Muff
    Certifications/Compliance ISO 9001 / 2000
    Color Metallic blue
    Dielectric Yes
    Noise Reduction Rating (NRR) 20dB
  • Description & Specifications
Related Products
Product Content Feedback