Howard Leight™ Clarity™ C3 Ear Muffs
Improve worker safety by blocking noise, while voice and signal frequencies can be heard more naturally
Manufacturer: Honeywell Safety Products 1011147
 View more versions of this product
Catalog No. 19-161-275
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabricationPetrochemicalSteelMiningTransportationUtility/Waste Management
    Style Cap mount
    Product Type Ear Muff
    Color Metallic blue
    Dielectric Yes
    Material Plastic
    Noise Reduction Rating (NRR) 27dB
  • Description & Specifications
Related Products
Product Content Feedback