Learn More
Honeywell Safety Products™ Com-Fit™ Ear Plugs
Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-025001
View Documents
Honeywell Safety Products™
281403
N/A Medium Size; Orange; Uncorded This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description
Description
NRR: 26dB
Durable. Made from nontoxic, nonirritating silicone rubber. Feature a triple-flange design for optimal fit and comfort, regardless of ear canal size. Can be sterilized in alcohol or boiling water. Comes with device that makes insertion easy and reduces attenuation variability. Noise Reduction Rating (NRR): 26dB.
Order Info
Available corded and uncorded, both styles come one pair per pack and in three color-coded sizes to facilitate safety checks.