Howard Leight™ DeciGuard AB™ Ear Plugs
Manufacturer: Honeywell Safety Products 280250/DISC
 View more versions of this product
Catalog No. 19-003-159C
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Product Type Deciguard Ab™ Earplug
    Material PVC
    Style Corded
    Noise Reduction Range (NRR) 29dB
    Certifications/Compliance ANSI S3.19-1974
    For Use With (Application) Lightweight, high-density PVC Foam provides rapid expansion in the ear canal for increased comfort and fit, nontoxic and nonirritating
  • Description & Specifications
Related Products
Product Content Feedback