Learn More
Honeywell Safety Products™ Howard Leight™ Ear Plug Dispense Refills
Single-use shouldn't mean single choice. Choose from a range of single-Use earplugs that include the comfort features your employees deserve, plus the highest SLC80 ratings available. An economical choice for work situations that demand a high degree of comfort, frequent changes, or where hygiene presents a problem for re-use.
Manufacturer:  Honeywell Safety Products™ MM1D
 View more versions of this product
Catalog No. 19-063-071
Add to cart
Description
Description
  • Vibrant colours make the Laser Lite a highly visible earplug and allows at-a-glance determination of worker compliance.
  • Max Lite provides comfort for workers with smaller ear canals in a low pressure foam earplug.
  • The Max is a bell shaped earplug, offering users maximum in-ear comfort, along with high attenuation.
Specifications
Specifications
Safety
Coral/yellow
Multi Max Single-use Earplug
Polyurethane Foam
31
Pill
EN 352-2:1993, EN 24869-1:1993, NRR 31
Uncorded
Howard Light dispenser
24dB
500 Pack