Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
PIP™ Seamless Knit Polyester Glove
Click to view available options
Size:
X-Small
Small
Medium
Large
X-Large
2X-Large
Description
- Seamless knit polyester shell offers increased comfort, finger dexterity and breathability.
- Polyurethane coatings take on the surface properties of the glove liner providing a tactile grip in dry and slightly oily conditions, ideal for applications that require dexterity for precise handling.
- Knit wrist helps prevent dirt and debris from entering the glove.
- Breathable back for comfort.
- Pair packed for ease of distribution.
- Silicone free.
Specifications
ANSI Abrasion Rating | ANSI 3 |
Chlorinated | No |
Construction | Seamless Knit |
Coating Material | Polyurethane |
Color | Black |
Exterior Finish | Smooth |
Hand | Specific |
Material | Polyester |
Product Type | Seamless Knit Gloves |
Size | Medium |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction