Howard Leight™ LS-400 Leight™ Source Ear Plug Dispenser

For convenient access to ear plugs

Manufacturer: Honeywell Safety Products MAXLS4

 View more versions of this product

Catalog No. 19-161-460

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance EN 352-2:2002
    Color Coral
    Cord Type Uncorded
    For Use With (Application) Agriculture, chemical, construction, fire protection, forestry, government, law enforcement, manufacturing, medicalMilitary, mining, municipal services, oil and gas, pharmaceutical, Steel and Metals, transportation, utilities, welding
    For Use With (Equipment) For LS-400 dispenser
    Material Polyurethane Foam
    Noise Level 26dB
    Noise Reduction Range (NRR) 33
    Quantity 2000 Case
    Shape Bell
    Surface Smooth
    Product Type Max Single-use Earplug

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit