Learn More
Howard Leight™ LS-400 Leight™ Source Ear Plug Dispenser
For convenient access to ear plugs
Manufacturer:  Honeywell Safety Products LLLS4
 View more versions of this product
Catalog No. 19-161-462
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Laser Lite Single-use Earplug
Uncorded
For LS-400 dispenser
25dB
2000 Case
Smooth
Magenta/yellow
Agriculture, chemical, construction, fire protection, forestry, government, law enforcement, manufacturing, medicalMilitary, mining, municipal services, oil and gas, pharmaceutical, Steel and Metals, transportation, utilities, welding
Polyurethane Foam
32
T-shaped