Learn More
Howard Leight™ Max-Lite™ Ear Plugs Get IT Item
NRR: 30dB
Manufacturer:  Honeywell Safety Products LPF1
 View more versions of this product
Catalog No. 19-045-765
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Green
Howard Leight™ Max Lite™ Earplug
T
30dB
Agriculture, chemical, construction, fire protection, law enforcement, medical, municipal services, transportation, welding
Polyurethane Foam
Uncorded
ANSI S3.19-1974