Learn More
Honeywell Safety Products™ MultiMax™ Ear Plugs
NRR: 31dB
Manufacturer:  Honeywell Safety Products™ MM1
 View more versions of this product
Catalog No. 19-003-331
Add to cart
Description
Description
  • Two-in-one ear plug lets you choose the tip that fits best
  • Unique design provides custom comfort; eliminates the need to stock several sizes
  • Smooth, soil-resistant surface in high-visibility colors
  • Uncorded
Order Info
Packaged in polybags or biodegradable, repulpable paper bags.
Compliance
Attenuation tested in accordance with ANSI™ S3.19-1974.
Specifications
Specifications
ANSI S3.19-1974
Smooth, soil resistant closed cell-foam skin prevents dirt build-up
Polyurethane Foam
Packed in Polybags
Coral, Yellow
Howard Leight™ Multimax™ Earplug
31 dB
Pill