Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Honeywell Safety Products™ MultiMax™ Ear Plugs

NRR: 31dB

$112.00

Specifications

Certifications/Compliance ANSI S3.19-1974
Color Coral, Yellow
Product Type Howard Leight™ Multimax™ Earplug
For Use With (Application) Smooth, soil resistant closed cell-foam skin prevents dirt build-up
Material Polyurethane Foam
View More Specs

Products 2
Catalog Number Mfr. No. Packaging Type Price Quantity & Availability  
Catalog Number Mfr. No. Packaging Type Price Quantity & Availability  
19-003-331
View Documents
Honeywell Safety Products™
MM1
Packed in Polybags
Pack of 200 for $112.00
Case of 10 PK for $1,002.50
Add to cart
 
19-037-408
View Documents
Honeywell Safety Products™
MM1PB
Packed in Repulpable Paper Bags This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
N/A
Description

Description

  • Two-in-one ear plug lets you choose the tip that fits best
  • Unique design provides custom comfort; eliminates the need to stock several sizes
  • Smooth, soil-resistant surface in high-visibility colors
  • Uncorded

Order Info

Packaged in polybags or biodegradable, repulpable paper bags.

Compliance

Attenuation tested in accordance with ANSI™ S3.19-1974.

Specifications

Specifications

ANSI S3.19-1974
Howard Leight™ Multimax™ Earplug
Polyurethane Foam
Pill
Coral, Yellow
Smooth, soil resistant closed cell-foam skin prevents dirt build-up
31 dB
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit