Howard Leight™ MultiMax™ Ear Plugs

NRR: 31dB

Manufacturer: Honeywell Safety Products MM1PB

 View more versions of this product

Catalog No. 19-037-408

  Check Availability
Add to Cart

    Specifications

    Product Type Howard Leight™ Multimax™ Earplug
    Material Polyurethane Foam
    Shape Pill
    Style Packed in Repulpable Paper Bags
    Noise Reduction Range (NRR) 31dB
    Certifications/Compliance ANSI S3.19-1974
    Color Coral/yellow
    For Use With (Application) Smooth, soil resistant closed cell-foam skin prevents dirt build-up

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit