Learn More
Honeywell Safety Products™ Quiet Bands™ HYG
NRR: 27 to 23dB
Manufacturer:  Honeywell Safety Products™ QB1HYG
 View more versions of this product
Catalog No. 19-033-674
Add to cart
Description
Description
  • Suspended pod design prevents contact with unsanitary surfaces
  • Pads made of soil-resistant, high-attenuating foam that can be easily cleaned and reused
  • Extra-wide ergonomic bands, each packaged with an extra pair of pods
QB1HYG
  • Combines the Quiet ear plug with the Quiet Band
  • Comfortable, high-attenuating protector for inner-aural use
  • NRR: 27dB
QB2HYG
  • Softly styled foam pads fit easily inside ear canal
  • For supra-aural protection; ideal for intermittent use
  • NRR: 25dB
QB3HYG
  • Semi-aural pads seal off ear canal without insertion
  • Comfortable, easy fit for moderate or intermittent hearing protection
  • NRR: 23dB
Specifications
Specifications
Ear Plug
27 dB
ANSI S3.19-1974, ANSI S12.6-1997, CSA Z94.2-1994, EN 352-2:1993, NRR 27
For inner-aural use
Polypropylene, Vinyl
Bell
Yellow, Orange
Pair of replacement pods
SDS
Documents
Accessories