Howard Leight™ Quiet™ Ear Plugs

NRR: 26dB

Manufacturer: Honeywell Safety Products QD1

 View more versions of this product

Catalog No. 18-777

  Check Availability
Add to Cart

    Specifications

    Material Vinyl
    Certifications/Compliance ANSI S12.6-1984
    Product Type Quiet™ Multiple-Use Earplug
    Shape Bell
    Style Uncorded
    Noise Reduction Range (NRR) 26dB
    Color Orange
    For Use With (Application) Multiple-use foam ear plugs with patented, no-roll design are easy to handle and fit

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit