Howard Leight™ SmartFit™ Ear Plugs

NRR: 25dB

Manufacturer: Honeywell Safety Products SMF30BU

 View more versions of this product

Catalog No. 19-130-3473

  Check Availability
Add to Cart

    Specifications

    Product Type Howard Leight™ Smartfit™ Earplug
    Shape 3 Flange
    Style Detachable Nylon cord
    Color Blue
    For Use With (Application) For pulp and paper, or tobacco processing industries
    Material TPE
    Noise Reduction Range (NRR) 25dB

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit