Howard Leight™ Viking™ Ear Muffs
Wear over/behind-the-head or under-the-chin
Manufacturer: Honeywell Safety Products 1010926
 View more versions of this product
Catalog No. 19-161-588
  Check Availability
Add to Cart
    Specifications
    Certifications/Compliance ANSI S.3.19-1974
    Color Blue
    Dielectric Yes
    For Use With (Application) Agriculture and farming, building construction, forestry, general contracting, heavy construction, landscaping, lumber or wood products, metal fabrication
    Petrochemical
    Steel
    Mining
    Transportation
    Utility/Waste Management
    Noise Reduction Rating (NRR) 27dB
    Style Viking 2 Earmuffs
    Product Type Ear Muff
  • Description & Specifications
Related Products
Product Content Feedback