Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA3 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548830

Catalog No. PA548830

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with zebrafish, bovine, chicken and human based on sequence homology.

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
Specifications

Specifications

Hoxa3
Polyclonal
PBS with 0.09% sodium azide
P02831
Homeobox protein Hox-A3, Homeobox protein Hox-15, Homeobox protein MO-10, Hoxa3, Hox-15, Hoxa-3
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
15400
Western Blot
Unconjugated
Hoxa3
Liquid
HOXA3
KLH conjugated synthetic peptide between 416-443 amino acids from the C-terminal region of mouse Hoxa3.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit