Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXA4 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5115377
Description
Antibody detects endogenous levels of total HOXA4.
In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
Specifications
HOXA4 | |
Polyclonal | |
Unconjugated | |
HOXA4 | |
AV206827; Dfd-like protein; homeo box A4; homeobox A4; homeobox protein Hox-1.4; Homeobox protein Hox-1D; homeobox protein Hox-A4; homeobox protein Hox-A4-like; homeobox protein MH-3; HOX1; Hox-1.4; Hox-1.4-like protein; Hox-14; HOX1D; Hoxa4; Hoxa-4; LOC100912525 | |
Rabbit | |
Affinity chromatography | |
RUO | |
3201 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
Q00056 | |
HOXA4 | |
A synthesized peptide derived from human HOXA4(Accession Q00056), corresponding to amino acid residues E81-H131. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction