Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXA4 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA550510
Description
The antibody detects endogenous levels of total HOXA4 protein.
HOXA4 Polyclonal antibody specifically detects HOXA4 in Human samples. It is validated for Immunohistochemistry (Paraffin).Specifications
HOXA4 | |
Polyclonal | |
Unconjugated | |
HOXA4 | |
AV206827; Dfd-like protein; homeo box A4; homeobox A4; homeobox protein Hox-1.4; Homeobox protein Hox-1D; homeobox protein Hox-A4; homeobox protein Hox-A4-like; homeobox protein MH-3; HOX1; Hox-1.4; Hox-1.4-like protein; Hox-14; HOX1D; Hoxa4; Hoxa-4; LOC100912525 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3201 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
0.6 mg/mL | |
PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q00056 | |
HOXA4 | |
Synthetic peptide corresponding to a region derived from internal residues of human homeobox A4 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction