Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXA4 Rabbit anti-Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548714

Catalog No. PA548714

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

In vertebrates, the genes encoding the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development. This gene is part of the A cluster on chromosome 7 and encodes a DNA-binding transcription factor which may regulate gene expression, morphogenesis, and differentiation.
Specifications

Specifications

Hoxa4
Polyclonal
PBS with 0.09% sodium azide
P06798
Homeobox protein Hox-A4, Homeobox protein Hox-14, Homeobox protein MH-3, Hoxa4, Hox-14, Hoxa-4
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
15401
Western Blot
Unconjugated
Hoxa4
Liquid
HOXA4
KLH conjugated synthetic peptide between 51-79 amino acids from the N-terminal region of mouse Hoxa4.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit