Learn More
HOXA9 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 185011AP
Catalog No. 50-173-2168
Add to cart
Description
Description
HOXA9 is the class of transcription factors called homeobox genes are found in clusters named A, B, C, and D on four separate chromosomes. Expression of these proteins is spatially and temporally regulated during embryonic development.
Specifications
Specifications
HOXA9
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P09631, P31269
ABD B, homeobox A9, Homeobox protein Hox 1G, Homeobox protein Hox A9, HOX1, HOX1.7, HOX1G, HOXA9
Rabbit
IgG
150 μL
-20°C
Primary
15405, 297099, 3205
Immunoprecipitation, Western Blot
Unconjugated
HOXA9
Liquid
HOXA9
HOXA9 Fusion Protein Ag13350
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse, Rat