Learn More
HOXB1 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 187321AP
Catalog No. 50-173-2216
Add to cart
Description
Description
18732-1-AP is a rabbit polyclonal antibody raised against a peptide mapping within an internal region of human HOXB1.
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis. Acts on the anterior body structures.
Specifications
Specifications
HOXB1
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P14653, P17919
homeobox B1, Homeobox protein Hox 2I, Homeobox protein Hox B1, Hox 2.9, HOX2, HOX2I, HOXB1
Rabbit
IgG
150 μL
-20°C
Primary
15407, 3211
Western Blot
Unconjugated
HOXB1
Liquid
HOXB1
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse