Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXB13 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518603
Description
This antibody is tested in Peptide ELISA: antibody detection limit dilution 128,000.
HOXB13 encodes a transcription factor that belongs to the homeobox gene family. Genes of this family are highly conserved among vertebrates and essential for vertebrate embryonic development. This gene has been implicated to play a role in fetal skin development and cutaneous regeneration. In mice, a similar gene was shown to exhibit temporal and spatial colinearity in the main body axis of the embryo, but was not expressed in the secondary axes, which suggests functions in body patterning along the axis. This gene and other HOXB genes form a gene cluster at chromosome the 17q21-22 region.
Specifications
| HOXB13 | |
| Polyclonal | |
| Unconjugated | |
| HOXB13 | |
| homeo box B13; homeobox B13; homeobox protein Hox-B13; HOXA13; HOXB13; PSGD | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 10481 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| Q92826 | |
| HOXB13 | |
| Synthetic peptide sequence (KKVLAKVKNSATP) corresponding to the C-terminus amino acids of HOXB13 (aa 272-284). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction