Learn More
HOXB13 Sheep anti-Human, Polyclonal, R&D Systems™
Sheep Polyclonal Antibody
Manufacturer:  R&D Systems AF8156SP
Catalog No. AF8156SP
Add to cart
This item is not returnable. View return policy
Description
Description
HOXB13 Polyclonal antibody AF8156SP specifically detects HOXB13 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry, Western Blotting
Specifications
Specifications
HOXB13
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL, Immunocytochemistry 5-15 ug/mL
Q92826
homeo box B13, homeobox B13, homeobox protein Hox-B13, PSGD
Sheep
IgG
25 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human HOXB13 in direct ELISAs and Western blots.
Human
Immunocytochemistry, Immunohistochemistry, Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
HOXB13
E. coli-derived human HOXB13 Met1-Gln102 Accession # Q92826
Antigen Affinity-purified
RUO
Primary
10481
Reconstitute at 0.2 mg/mL in sterile PBS.