Learn More
HOXB4 Mouse anti-Human, Alexa Fluor 700, Clone: 3A2F2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237257AF700
Catalog No. NBP237257X
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
HOXB4
3A2F2
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
HOXB4
Purified recombinant fragment of human HOXB4 (AA: 16-251) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Alexa Fluor 700
50mM Sodium Borate with 0.05% Sodium Azide
homeo box 2F, homeo box B4, homeobox B4, Homeobox protein Hox-2.6, Homeobox protein Hox-2F, homeobox protein Hox-B4, HOX-2.6, HOX2FHOX2
Mouse
IgG2a
0.1 ml
Primary
3214