Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXB4 Mouse anti-Human, Biotin, Clone: 3A2F2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237257B

Catalog No. NBP237257B

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Specifications

Specifications

HOXB4
3A2F2
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
HOXB4
Purified recombinant fragment of human HOXB4 (AA: 16-251) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Biotin
PBS with 0.05% Sodium Azide
homeo box 2F, homeo box B4, homeobox B4, Homeobox protein Hox-2.6, Homeobox protein Hox-2F, homeobox protein Hox-B4, HOX-2.6, HOX2FHOX2
Mouse
IgG2a
0.1 ml
Primary
3214
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit