Learn More
HOXB4 Mouse anti-Human, HRP, Clone: 3A2F2, Novus Biologicals™
Mouse Monoclonal Antibody
Manufacturer:  Novus BiologicalsSupplier Diversity Partner NBP237257H
Catalog No. NBP237257H
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Specifications
HOXB4
3A2F2
Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin
Purified
HOXB4
Purified recombinant fragment of human HOXB4 (AA: 16-251) expressed in E. Coli.
Protein A or G purified
Store at 4C in the dark.
Monoclonal
Human
ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
HRP
PBS with No Preservative
homeo box 2F, homeo box B4, homeobox B4, Homeobox protein Hox-2.6, Homeobox protein Hox-2F, homeobox protein Hox-B4, HOX-2.6, HOX2FHOX2
Mouse
IgG2a
0.1 ml
Primary
3214