Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXB7 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548725

Catalog No. PA548725

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

The homeobox genes encode transcription factors that regulate developmental processes such as body patterning and organogenesis. The mammalian HOX gene complex consists of 39 genes that are located on 4 linkage groups (clusters), dispersed over 4 chromosomes (HOXA, HOXB, HOXC and HOXD). HOXB7 is expressed in most maligt T-cell lines and CD4-positive peripheral blood cells, and is constitutively expressed in both melanoma primary lesions and cell lines. HOXB7 binds to and transactivates bFGF in melanoma cell lines but SkBr3 breast adenocarcinoma is negative for the expression of both HOXB7 and bFGF genes. HOXB7 may be a key activator of tumor-associated neoangiogenesis.
Specifications

Specifications

Hoxb7
Polyclonal
PBS with 0.09% sodium azide
P09629, P09024
Homeobox protein Hox-B7, Homeobox protein Hox-23, Homeobox protein MH-22B, Homeobox protein MuB1, Hoxb7, Hox-23, Hoxb-7
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
15415, 3217
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
HOXB7
Liquid
HOXB7
KLH conjugated synthetic peptide between 94-122 amino acids from the Central region of mouse Hoxb7.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit