Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXB8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567398
Description
Immunogen sequence: NNKDKFPSSKC EQEELEKQKL ERAPEAADEG DAQK Highest antigen sequence identity to the following orthologs - mouse 89%, rat 89%.
This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Increased expression of this gene is associated with colorectal cancer. Mice that have had the murine ortholog of this gene knocked out exhibit an excessive pathologic grooming behavior. This behavior is similar to the behavior of humans suffering from the obsessive-compulsive spectrum disorder trichotillomania.
Specifications
HOXB8 | |
Polyclonal | |
Unconjugated | |
HOXB8 | |
homeo box 2D; homeo box B8; homeobox B8; homeobox gene B8; homeobox protein Hox-2.4; Homeobox protein Hox-2D; homeobox protein Hox-B8; homeobox protein R1a; HOX2; Hox-2.4; Hox2.4 homeobox homolog; HOX2D; Hox2r1a; Hoxb8; Hoxb-8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
3218 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
P17481 | |
HOXB8 | |
Recombinant Human HOXB8. Recombinant protein control fragment (Product #RP-108062). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction