Learn More
HOXC10 Rabbit anti-Human, Polyclonal, Abnova™
Rabbit Polyclonal Antibody
Manufacturer:  Abnova Corporation PAB30144
Catalog No. 89-033-432
Add to cart
Description
Description
Rabbit polyclonal antibody raised against synthetic peptide of human HOXC10.
  • High specificity
  • High reactivity
Specifications
Specifications
HOXC10
Unconjugated
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (4-8 ug/mL) Western Blot (0.0625 ug/mL) The optimal working dilution should be determined by the end user.
NM_017409
HOX3I|MGC5259
Rabbit
Affinity purification
RUO
Primary
3226
Immunohistochemistry (PFA fixed), Western Blot
Rabbit polyclonal antibody raised against synthetic peptide of human HOXC10.
In PBS (2% sucrose, 0.09% sodium azide).
Liquid
HOXC10
A synthetic peptide corresponding to N-terminus of human HOXC10.
100 uL
Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Polyclonal
Human
Documents
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Cancel Submit
For Research Use Only (RUO)