Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXC11 Mouse anti-Human, Allophycocyanin/Cy7, Clone: OTI3E10, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270913APCCY7

Catalog No. N270913APC7


Add to cart

Description

Description

HOXC11 Monoclonal antibody specifically detects HOXC11 in Human samples. It is validated for Flow Cytometry.
Specifications

Specifications

HOXC11
OTI3E10
Flow Cytometry
Affinity Purified, Conjugated
HOXC11
Recombinant protein fragment corresponding to amino acids 1-304 of human HOXC11 (NP_055027) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark. Do not freeze.
Monoclonal
Human
Flow Cytometry
APC-Cyanine7
PBS with 0.05% Sodium Azide
homeo box C11, homeobox C11, Homeobox protein Hox-3H, homeobox protein Hox-C11, HOX3H, MGC4906
Mouse
IgG1
0.1 mL
Primary
3227.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit