Learn More
HOXC11 Rabbit anti-Human, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 221181AP
Catalog No. 50-173-3730
Add to cart
Description
Description
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene binds to a promoter element of the lactase-phlorizin hydrolase. It also may play a role in early intestinal development. An alternatively spliced variant encoding a shorter isoform has been described but its full-length nature has not been determined.
Specifications
Specifications
HOXC11
0.31 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O43248
homeobox C11, Homeobox protein Hox 3H, Homeobox protein Hox C11, HOX3H, HOXC11
Rabbit
IgG
150 μL
-20°C
Primary
3227
Western Blot
Unconjugated
HOXC11
Liquid
HOXC11
HOXC11 Fusion Protein Ag17806
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human