Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hoxc5 Polyclonal Antibody
Hoxc5 Polyclonal Antibody
Supplier: Thermo Scientific PA555421
Description
Hoxc5 Polyclonal Antibody for ICC/IF, IHC

Specifications
Hoxc5 | |
Polyclonal | |
Unconjugated | |
HOXC5 | |
AI181837; CP11; homeo box C5; homeobox C5; homeobox protein CP11; Homeobox protein Hox-3.4; homeobox protein Hox-3D; Homeobox protein Hox-6.2; homeobox protein Hox-C5; HOX3; Hox-3.4; Hox-34; HOX3D; Hox-6.2; HOXC5; Hoxc-5 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
3222 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Immunocytochemistry | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q00444 | |
HOXC5 | |
Recombinant protein corresponding to Human Hoxc5. Recombinant protein control fragment (Product #RP-102136). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction