Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hoxc5 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548909

Catalog No. PA548909

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

Predicted to react with zebrafish and xenopus based on sequence homology.

Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications

Specifications

Hoxc5
Polyclonal
PBS with 0.09% sodium azide
Q00444, P32043
Homeobox protein Hox-C5, Homeobox protein Hox-34, Homeobox protein Hox-62, Hoxc5, Hox-34, Hoxc-5
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
15424, 3222
Western Blot
Unconjugated
HOXC5
Liquid
HOXC5
KLH conjugated synthetic peptide between 140-167 amino acids from the C-terminal region of mouse Hoxc5.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit