Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ HOXC8 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA541629
Description
Peptide sequence: SHALVYGPGG SAPGFQHASH HVQDFFHHGT SGISNSGYQQ NPCSLSCHGD Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Sheep: 100%; Zebrafish: 77%.
This gene belongs to the homeobox family of genes. The homeobox genes encode a highly conserved family of transcription factors that play an important role in morphogenesis in all multicellular organisms. Mammals possess four similar homeobox gene clusters, HOXA, HOXB, HOXC and HOXD, which are located on different chromosomes and consist of 9 to 11 genes arranged in tandem. This gene is one of several homeobox HOXC genes located in a cluster on chromosome 12. The product of this gene may play a role in the regulation of cartilage differentiation. It could also be involved in chondrodysplasias or other cartilage disorders.
Specifications
HOXC8 | |
Polyclonal | |
Unconjugated | |
HOXC8 | |
D130011F21Rik; homeo box 3A; homeo box C8; homeobox C8; Homeobox gene C8; homeobox protein Hox-3.1; Homeobox protein Hox-3A; homeobox protein Hox-C8; Homeobox protein M31; homeobox protein R4; HOX3; Hox-3.1; Hox-3.1, mouse, homolog of; HOX3A; Hox3r4; HOXC8; Hoxc-8 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
3224 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P31273 | |
HOXC8 | |
Synthetic peptide directed towards the N-terminal of human HOXC8 (aa 35-84). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction