Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HOXD10 Rabbit anti-Human, Mouse, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Thermo Scientific PA548685
Description
This gene is a member of the Abd-B homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox D genes located on chromosome 2. The encoded nuclear protein functions as a sequence-specific transcription factor that is expressed in the developing limb buds and is involved in differentiation and limb development. Mutations in this gene have been associated with Wilm′s tumor and congenital vertical talus and/or a foot deformity resembling that seen in Charcot-Marie-Tooth disease.Specifications
Hoxd10 | |
Polyclonal | |
PBS with 0.09% sodium azide | |
P28358, P28359 | |
HOXD10 | |
KLH conjugated synthetic peptide between 201-230 amino acids from the Central region of mouse Hoxd10. | |
400 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
HOXD10 | |
Homeobox protein Hox-D10, Homeobox protein Hox-45, Homeobox protein Hox-53, Hoxd10, Hox-45, Hoxd-10 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
15430, 3236 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction