Learn More
HOXD12 Rabbit anti-Human, Mouse, Polyclonal, Proteintech Non-distribution product as customer accommodation.
Rabbit Polyclonal Antibody
Manufacturer:  Proteintech Group IncSupplier Diversity Partner 187351AP
Catalog No. 50-173-2218
Add to cart
Description
Description
The Hox proteins are a family of transcription factors that play a role in development and cellular differentiation by regulating downstream target genes. Specifically, the Hox proteins direct DNA-protein and protein-protein interactions that assist in determining the morphologic features associated with the anterior-posterior body axis. Hox proteins are involved in controlling axial patterning, leukemias and hereditary malformations. HoxD12 (homeobox D12), also known as HOX4H, is a 275 amino acid protein that localizes to the nucleus and contains one homeobox DNA-binding domain. One of several members of the homeobox superfamily, HoxD12 functions as a sequencespecific transcription factor that is important for the correct positioning of developing limb buds on the anterior-posterior axis.
Specifications
Specifications
HOXD12
0.13 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
P23812, P35452
homeobox D12, Homeobox protein Hox 4H, Homeobox protein Hox D12, HOX4H, HOXD12
Rabbit
IgG
150 μL
-20°C
Primary
15432, 3238
Western Blot
Unconjugated
HOXD12
Liquid
HOXD12
Peptide
Antigen Affinity Chromatography
RUO
Antibody
Polyclonal
Human, Mouse