Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXD13 Goat anti-Rat, Polyclonal, Novus Biologicals™

Goat Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP275091

Catalog No. NBP275091


Add to cart

Description

Description

HOXD13 Polyclonal antibody specifically detects HOXD13 in Rat samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

HOXD13
0.5 mg/ml
Western Blot 1:100-1:2000, Peptide ELISA
Purified
HOXD13
Peptide with sequence C-HGVGLQQNALKSSPH, from the internal region of the protein sequence according to NP_000514.2.
34.9 kDa
100 μg
Primary
3239.0
ELISA, Western Blot
Unconjugated
TBS (pH 7.3), 0.5% BSA with 0.02% Sodium Azide
BDSD, homeo box 4I, homeo box D13, homeobox D13, Homeobox protein Hox-4I, homeobox protein Hox-D13, HOX4IBDE, SPD
Goat
IgG
Antigen Affinity-purified
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Rat
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit