Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HOXD4 Mouse anti-Human, Alexa Fluor™ 405, Clone: OTI1E9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP270919AF405

Catalog No. N270919A405


Add to cart

Description

Description

HOXD4 Monoclonal antibody specifically detects HOXD4 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

HOXD4
OTI1E9
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
HOXD4
Full length recombinant protein of human HOXD4 (NP_055436) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
Alexa Fluor 405
50mM Sodium Borate with 0.05% Sodium Azide
HHO.C13, homeo box D4, homeobox D4, Homeobox protein HHO.C13, Homeobox protein Hox-4B, Homeobox protein Hox-5.1, homeobox protein Hox-D4, HOX4, Hox-4.2, Hox-4.2, mouse, homolog of homeo box X, HOX4BHOX-5.1
Mouse
IgG2a
0.1 mL
Primary
3233.0
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit