Learn More
HOXD4 Mouse anti-Human, Clone: OTI4E7, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF809488
Catalog No. 50-166-9531
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Sequence-specific transcription factor which is part of a developmental regulatory system that provides cells with specific positional identities on the anterior-posterior axis.
Specifications
Specifications
HOXD4
Monoclonal
Unconjugated
HOXD4
Lyophilized
HOXD4
Full length human recombit protein of human HOXD4 produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI4E7
PBS with 8% trehalose and no preservative; pH 7.3
P09016
HHO.C13, HOX-5.1, HOX4, HOX4B, Hox-4.2
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
3233